2-Methyl-1,3-cyclohexanedione
Supplier: Cayman Chemical
Synonyms: 2-Methylcyclohexane-1,3-dione
10007138-1 10007138-10 10007138-5
CAYM10007138-1EA 28 EUR
CAYM10007138-1 CAYM10007138-10 CAYM10007138-5
2-Methyl-1,3-cyclohexanedione
2-Methyl-1,3-cyclohexanedione
Formula: C₇H₁₀O₂
MW: 126.16 g/mol
Boiling Pt: 100…102 °C (8 mmHg)
Melting Pt: 206…209 °C
Density: 1.313 g/cm³ (–150.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001587
CAS Number: 1193-55-1
EINECS: 214-773-9