3,5-Dimethyl-1,2-cyclopentanedione, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3,5-Dimethylcyclopentane-1,2-dione
M-Clarity™ quality level = MQ300
W326909-100G-K
W326909-1KG-K
W326909-25G-K
SIALW326909-100G-KEA
540
EUR
SIALW326909-100G-K
SIALW326909-1KG-K
SIALW326909-25G-K
3,5-Dimethyl-1,2-cyclopentanedione, Sigma-Aldrich®
3,5-Dimethyl-1,2-cyclopentanedione
Formula:
C₇H₁₀O₂ MW: 126.16 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00143076 CAS Number: 13494-07-0 EINECS: 236-811-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...