Cyclobutylméthanol 99%
Supplier: Thermo Scientific
Warning
Synonyms: Cyclobutanemethanol, Cyclobutylmethanol, Hydroxymethylcyclobutane
182230050 182230250
ACRO182230050EA 62 EUR
ACRO182230050 ACRO182230250
Cyclobutylméthanol 99%
Cyclobutylméthanol
Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 143…144 °C
Density: 0.91 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00001330
CAS Number: 4415-82-1
EINECS: 224-575-4
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4470 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %