Cyclopropylméthanol 99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol
296820250 296820010 296821000 296822500
ACRO296820250EA 49.7 EUR
ACRO296820250 ACRO296820010 ACRO296821000 ACRO296822500
Cyclopropylméthanol 99%
Cyclopropylméthanol
Formula: C₄H₈O
Boiling Pt: 123…124 °C (738 mmHg)
Melting Pt: –60 °C
Density: 0.89 g/cm³
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00001309
CAS Number: 2516-33-8
EINECS: 219-735-5
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥98.5 %