Cyclopropylméthanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cyclopropyl carbinol, Cyclopropanemethanol
M-Clarity™ quality level = MQ100
482099-1L 482099-250ML 482099-25ML
SIAL482099-1LEA 597 EUR
SIAL482099-1L SIAL482099-250ML SIAL482099-25ML
Cyclopropylméthanol, Sigma-Aldrich®
Cyclopropylméthanol
Formula: C₄H₈O
Boiling Pt: 123 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.904 g/cm³ (20 °C)
MDL Number: MFCD00001309
CAS Number: 2516-33-8
EINECS: 219-735-5
UN: 1987
ADR: 3,III