1-Methylcyclohexanol, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

M38214-100G M38214-25G
SIALM38214-100GEA 258 EUR
SIALM38214-100G SIALM38214-25G
1-Methylcyclohexanol, Sigma-Aldrich®
1-Methylcyclohexanol
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: 26 °C
Density: 0.9282 g/cm³ (20 °C)
MDL Number: MFCD00003857
CAS Number: 590-67-0
EINECS: 209-688-9
UN: 2617
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR