Methylenecyclohexane, Sigma-Aldrich®

Supplier: Merck

111015-1G 111015-5G
SIAL111015-1GEA 85.7 EUR
SIAL111015-1G SIAL111015-5G
Methylenecyclohexane, Sigma-Aldrich®
Methylenecyclohexane
Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 103
Melting Pt: –106.7 °C
Density: 0.8074 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001661
CAS Number: 1192-37-6
EINECS: 214-752-4
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR