2-Cyclopenten-1-one, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Cyclopentenone, Cyclopenten-2-one, Cyclopent-2-enone
M-Clarity™ quality level = MQ200
C112909-100G C112909-25G C112909-5G
SIALC112909-100GEA 574 EUR
SIALC112909-100G SIALC112909-25G SIALC112909-5G
2-Cyclopenten-1-one, Sigma-Aldrich®
2-Cyclopenten-1-one
Formula: C₅H₆O
MW: 82.1 g/mol
Boiling Pt: 64 °C (1013 hPa)
Density: 0.989 g/cm³ (15 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001401
CAS Number: 930-30-3
EINECS: 213-213-0
UN: 1224
ADR: 3,III