2-Methyl-2-cyclopenten-1-one, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
304190-1G 304190-5G
SIAL304190-1GEA 81.2 EUR
SIAL304190-1G SIAL304190-5G
2-Methyl-2-cyclopenten-1-one, Sigma-Aldrich®
2-Methyl-2-cyclopenten-1-one
Formula: C₆H₈O
MW: 96.13 g/mol
Boiling Pt: 158 °C (1013 hPa)
Density: 0.9772 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00012275
CAS Number: 1120-73-6
UN: 1224
ADR: 3,III