3-Methyl-2-cyclohexen-1-one, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Methyl-2-cyclohexenone, 3-Methylcyclohex-2-enone
M-Clarity™ quality level = MQ100
M39105-10G M39105-50G
SIALM39105-10GEA 95.7 EUR
SIALM39105-10G SIALM39105-50G
3-Methyl-2-cyclohexen-1-one, Sigma-Aldrich®
3-Methyl-2-cyclohexen-1-one
Formula: C₇H₁₀O
MW: 110.16 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 198…199 °C
Density: 0.9791 g/cm³ (10 °C)
MDL Number: MFCD00001581
CAS Number: 1193-18-6
EINECS: 214-769-7
UN: 1224
ADR: 3,III