Hexamethylcyclotrisiloxane 98%
Supplier: Thermo Scientific
Danger
Synonyms: 2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-trioxatrisilinane
216460250 216461000 216465000
ACRO216460250EA 30 EUR
ACRO216460250 ACRO216461000 ACRO216465000
Hexamethylcyclotrisiloxane 98%
Hexamethylcyclotrisiloxane
Formula: C₆H₁₈O₃Si₃
MW: 222.46 g/mol
Boiling Pt: 134 °C (760 mmHg)
Melting Pt: 63…67 °C
Density: 1,12 g/cm³ (20 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00005943
CAS Number: 541-05-9
EINECS: 208-765-4
UN: 1325
ADR: 4.1,III
Order Now
Specification Test Results
Appearance Colorless to white Crystalline chunks
Infrared spectrum Conforms
Melting point 63 to 67 °C
GC ≥97.5 %