Hexamethylcyclotrisiloxane ≥97%
Supplier: Thermo Scientific
Danger
Synonyms: 2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-trioxatrisilinane
B23028.14 B23028.22 B23028.36
B23028.14EA 32 EUR
B23028.14 B23028.22 B23028.36
Hexamethylcyclotrisiloxane ≥97%
Hexamethylcyclotrisiloxane
Purity: 97% CAS No.: 541-05-9 Molecular Formula: C6H18O3Si3 Formula Weight: 222.47 Flash Point: 35°(95°F) Melting Point: 64-66° Boiling Point: 134° Beilstein Reference: 1766829 UN No.: 1325 MDL No.: MFCD00005943
Formula: C₆H₁₈O₃Si₃
MW: 222.46 g/mol
Boiling Pt: 134 °C
Melting Pt: 64…66 °C
Density: 1.12 g/cm³ (20 °C)
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005943
CAS Number: 541-05-9
EINECS: 208-765-4
UN: 1325
ADR: 4.1,II