Hexamethylcyclotrisiloxane, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,2,4,4,6,6-Hexamethyl-1,3,5,2,4,6-trioxatrisilinane
M-Clarity™ quality level = MQ100
235687-25G 235687-100G
SIAL235687-25GEA 42.4 EUR
SIAL235687-25G SIAL235687-100G
Hexamethylcyclotrisiloxane, Sigma-Aldrich®
Hexamethylcyclotrisiloxane
Formula: C₆H₁₈O₃Si₃
MW: 222.46 g/mol
Boiling Pt: 134 °C (1013 hPa)
Melting Pt: 64.5 °C
Density: 1.12 g/cm³ (20 °C)
MDL Number: MFCD00005943
CAS Number: 541-05-9
EINECS: 208-765-4
UN: 1325
ADR: 4.1,II