2-Thiobarbituric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
4,6-Dihydroxy-2-mercaptopyrimidine
M-Clarity™ quality level = MQ100
T5500-100G
T5500-250G
T5500-25G
T5500-500G
SIALT5500-100GEA
250
EUR
SIALT5500-100G
SIALT5500-250G
SIALT5500-25G
SIALT5500-500G
2-Thiobarbituric acid, Sigma-Aldrich®
2-Thiobarbituric acid
Formula:
C₄H₄N₂O₂S MW: 144.15 g/mol Boiling Pt: 419 °C (1013 hPa) Melting Pt: 235 °C Density: 1.661 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006674 CAS Number: 504-17-6 EINECS: 207-985-8 |