Barbituric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
M-Clarity™ quality level = MQ200
185698-100G
185698-25G
185698-500G
SIAL185698-100GEA
65.2
EUR
SIAL185698-100G
SIAL185698-25G
SIAL185698-500G
Barbituric acid, Sigma-Aldrich®
Barbituric acid
Formula:
C₄H₄N₂O₃ Boiling Pt: 441 °C (1013 hPa) Melting Pt: 250…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006666 CAS Number: 67-52-7 EINECS: 200-658-0 Merck Index: 13,00966 |