Piperazine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
80621-100G
80621-25G
80621-500G
SIAL80621-100GEA
209
EUR
SIAL80621-100G
SIAL80621-25G
SIAL80621-500G
Piperazine, Sigma-Aldrich®
Piperazine
Formula:
C₄H₁₀N₂ Boiling Pt: 146 °C (1013 hPa) Melting Pt: 44…46 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005953 CAS Number: 110-85-0 EINECS: 203-808-3 UN: 2579 ADR: 8,III Merck Index: 13,07545 |