Piperazine, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
P45907-100G P45907-500G P45907-5G P45907-1KG
SIALP45907-100GEA 33.5 EUR
SIALP45907-100G SIALP45907-500G SIALP45907-5G SIALP45907-1KG
Piperazine, Sigma-Aldrich®
Piperazine
Formula: C₄H₁₀N₂
MW: 86.14 g/mol
Boiling Pt: 146
Melting Pt: 44…46 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 65
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8 III
Merck Index: 13,07545