1-Phenylpiperazine, Sigma-Aldrich®
Supplier: Merck
Synonyms: N-Phenylpiperazine
M-Clarity™ quality level = MQ100
P30004-100G P30004-25G
SIALP30004-100GEA 119 EUR
SIALP30004-100G SIALP30004-25G
1-Phenylpiperazine, Sigma-Aldrich®
1-Phenylpiperazine
Formula: C₁₀H₁₄N₂
MW: 162.23 g/mol
Boiling Pt: 286 °C (1013 hPa)
Melting Pt: 17…19 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005957
CAS Number: 92-54-6
EINECS: 202-165-6
UN: 2927
ADR: 6.1,II