2,7-Dibromofluorene, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
2,7-Dibromo-9H-fluorene
M-Clarity™ quality level = MQ100
342297-25G
342297-5G
SIAL342297-25GEA
79.8
EUR
SIAL342297-25G
SIAL342297-5G
2,7-Dibromofluorene, Sigma-Aldrich®
2,7-Dibromofluorene
Formula:
C₁₃H₈Br₂ MW: 324.01 g/mol |
MDL Number:
MFCD00019048 CAS Number: 16433-88-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...