9,10-Anthraquinone 98%
Supplier: Thermo Scientific
Danger
Synonyms: Anthracene-9,10-dione, Anthraquinone
104930010 104932500 104930500
104930010.EA 102 EUR
104930010. 104932500. 104930500.
9,10-Anthraquinone 98%
9,10-Anthraquinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 379…381 °C (760 mmHg)
Melting Pt: 283…288 °C
Density: 1,419 to 1.438 g/cm³ (20 °C)
Flash Pt: 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692
Order Now
Specification Test Results
Appearance Yellow-green to tan to khaki Powder
Infrared spectrum Conforms
Melting point 283 to 288 °C
HPLC ≥97.5 %