9,10-Anthraquinone, Sigma-Aldrich®
Supplier: Merck
Synonyms: Anthracene-9,10-dione, Anthraquinone
M-Clarity™ quality level = MQ200
A90004-250G A90004-50G
SIALA90004-250GEA 79.2 EUR
SIALA90004-250G SIALA90004-50G
9,10-Anthraquinone, Sigma-Aldrich®
9,10-Anthraquinone
Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 165…169 °C
Density: 1.419…1.438 g/cm³ (20 °C)
Flash Pt: 185 °C
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692