1,4-Dihydroxy-9,10-anthraquinone 95%
Supplier: Thermo Scientific
Synonyms:
1,4-Dihydroxyanthracene-9,10-dione, Quinizarin, 1,4-Dihydroxyanthraquinone
A11010.0B
A11010.30
A11010.0BEA
127
EUR
A11010.0B
A11010.30
1,4-Dihydroxy-9,10-anthraquinone 95%
1,4-Dihydroxy-9,10-anthraquinone
Purity: 95%
CAS No.: 81-64-1
Molecular Formula: C14H8O4
Formula Weight: 240.21
Flash Point: 222°(431°F)
Melting Point: 195-198°
Boiling Point: ca 450°
Beilstein Reference: 1914036
Merck Reference: 14,8064
MDL No.: MFCD00001209
Formula:
C₁₄H₈O₄ MW: 240.22 g/mol Boiling Pt: ∼450 °C Melting Pt: 195…198 °C Density: 1.278 g/cm³ (20 °C) Flash Pt: 222 °C (431 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001209 CAS Number: 81-64-1 EINECS: 201-368-7 Merck Index: 14,8064 |