Dibenzofuran, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Dibenzo[b,d]furan
M-Clarity™ quality level = MQ100
236373-50G
236373-10G
SIAL236373-50GEA
166
EUR
SIAL236373-50G
SIAL236373-10G
Dibenzofuran, Sigma-Aldrich®
Dibenzofuran
Formula:
C₁₂H₈O Boiling Pt: 285 °C (1013 hPa) Melting Pt: 79…82 °C Density: 0.966 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004968 CAS Number: 132-64-9 EINECS: 205-071-3 |