Fluorescein disodium salt, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
M-Clarity™ quality level = MQ200
46960-100G-F 46960-25G-F 46960-500G-F
SIAL46960-100G-FEA 91.7 EUR
SIAL46960-100G-F SIAL46960-25G-F SIAL46960-500G-F
Fluorescein disodium salt, Sigma-Aldrich®
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
MW: 376.28 g/mol
Melting Pt: > 300 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 13,04185
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM