Propylene carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester
M-Clarity™ quality level = MQ100
310328-100ML 310328-1L 310328-2L 310328-500ML
SIAL310328-100MLEA 78.8 EUR
SIAL310328-100ML SIAL310328-1L SIAL310328-2L SIAL310328-500ML
Propylene carbonate, Sigma-Aldrich®
Propylene carbonate
Formula: C₄H₆O₃
MW: 102.09 g/mol
Boiling Pt: 241…242 °C (1013 hPa)
Melting Pt: –48.8 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 132 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005385
CAS Number: 108-32-7
EINECS: 203-572-1