5-(Hydroxymethyl)-2-furaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 5-Hydroxymethyl-2-furancarbaldehyde, 5-(hydroxymethyl)furan-2-carbaldehyde, 5-(Hydroxymethyl)furfural, 5-Hydroxymethylfurfural
M-Clarity™ quality level = MQ300
W501808-100G-K W501808-1KG-K W501808-25G-K W501808-1G-K
SIALW501808-100G-KEA 1220 EUR
SIALW501808-100G-K SIALW501808-1KG-K SIALW501808-25G-K SIALW501808-1G-K
5-(Hydroxymethyl)-2-furaldehyde, Sigma-Aldrich®
5-(Hydroxymethyl)-2-furaldehyde
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 292
Melting Pt: 27…30 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00003234
CAS Number: 67-47-0
EINECS: 200-654-9
Merck Index: 13,04857