2-Ethylfuran ≥98%
Supplier: Thermo Scientific
|
Danger
|
A15520.14EA
140
EUR
A15520.14
A15520.22
A15520.06
2-Ethylfuran ≥98%
2-Ethylfuran
Formula:
C₆H₈O MW: 96,13 g/mol Boiling Pt: 92…93 °C Density: 0,912 Flash Pt: –2 °C (28 °F) Storage Temperature: Ambient |
UN:
3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...