2-Ethylfuran, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
SIALW367303-25G-KEA
108
EUR
SIALW367303-25G-K
SIALW367303-100G-K
2-Ethylfuran, Sigma-Aldrich®
2-Ethylfuran
Formula:
C₆H₈O MW: 96,13 g/mol Boiling Pt: 92 °C (1013 hPa) Density: 0,912 g/cm³ (13 °C) |
UN:
3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...