1,1,3,3-Tetramethylguanidine
Supplier: Thermo Scientific
Danger
Synonyms: N,N,N',N'-Tetramethylguanidine
SB01651.1 SB01651.10 SB01651.50
MAYBSB01651.1EA 0 EUR
MAYBSB01651.1 MAYBSB01651.10 MAYBSB01651.50
1,1,3,3-Tetramethylguanidine
1,1,3,3-Tetramethylguanidine
Formula: C₅H₁₃N₃
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8,II