1,1,3,3-Tetramethylguanidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N,N,N',N'-Tetramethylguanidine
M-Clarity™ quality level = MQ200
690953-10KG 690953-1KG
SIAL690953-10KGEA 3380 EUR
SIAL690953-10KG SIAL690953-1KG
1,1,3,3-Tetramethylguanidine, Sigma-Aldrich®
1,1,3,3-Tetramethylguanidine
Formula: C₅H₁₃N₃
Boiling Pt: 160 °C (1013 hPa)
Melting Pt: –30 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00008323
CAS Number: 80-70-6
EINECS: 201-302-7
UN: 2920
ADR: 8,II