Molybdic acid (≥85% MoO₃)
Supplier: Thermo Fisher Scientific
Warning
A10537.0E A10537.22 A10537.36
A10537.0EEA 760 EUR
A10537.0E A10537.22 A10537.36
Molybdic acid (≥85% MoO₃)
Molybdic acid
CAS No.: 7782-91-4 MDL No.: MFCD00036287
Formula: H₂MoO₄
Boiling Pt: ∼300 °C (1013 hPa)
Melting Pt: 300 °C
Density: 3.1
Storage Temperature: Ambient
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262