di-Lithium tetraborate 99,995 Trace metals basis, Sigma-Aldrich®
Supplier: Merck
Synonyms: Lithium tetraborate, Dilithium tetraborate
MQ100
254304-100G 254304-25G
SIAL254304-100GEA 247 EUR
SIAL254304-100G SIAL254304-25G
di-Lithium tetraborate 99,995 Trace metals basis, Sigma-Aldrich®
di-Lithium tetraborate
Formula: Li₂B₄O₇
MW: 169.12 g/mol
Melting Pt: 918
Density: 1.4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011083
CAS Number: 12007-60-2
EINECS: 234-514-3
Merck Index: 14,05524