Lithium tetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
244767-10G
244767-50G
SIAL244767-10GEA
66
EUR
SIAL244767-10G
SIAL244767-50G
Lithium tetrafluoroborate, Sigma-Aldrich®
Lithium tetrafluoroborate
Formula:
BF₄Li MW: 93.74 g/mol Density: 0.852 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011087 CAS Number: 14283-07-9 EINECS: 238-178-9 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...