Lithium tetrafluoroborate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
451622-25G 451622-5G
SIAL451622-25GEA 631 EUR
SIAL451622-25G SIAL451622-5G
Lithium tetrafluoroborate, Sigma-Aldrich®
Lithium tetrafluoroborate
Formula: BF₄Li
Density: 0.852 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011087
CAS Number: 14283-07-9
EINECS: 238-178-9
UN: 3260
ADR: 8,III