Potassium tetrahydroborate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Potassium tetrahydridoborate, Potassium borohydride
M-Clarity™ quality level = MQ200
438472-25G 438472-5G
SIAL438472-25GEA 308 EUR
SIAL438472-25G SIAL438472-5G
Potassium tetrahydroborate, Sigma-Aldrich®
Potassium tetrahydroborate
Formula: H₄BK
MW: 53.94 g/mol
Melting Pt: 500 °C
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3,I
Merck Index: 12,07777