Potassium tetrahydroborate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Potassium tetrahydridoborate, Potassium borohydride
M-Clarity™ quality level = MQ200
455571-100G 455571-500G
SIAL455571-100GEA 85.9 EUR
SIAL455571-100G SIAL455571-500G
Potassium tetrahydroborate, Sigma-Aldrich®
Potassium tetrahydroborate
Formula: H₄BK
Melting Pt: 500 °C
Density: 1.17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011396
CAS Number: 13762-51-1
EINECS: 237-360-5
UN: 1870
ADR: 4.3,I
Merck Index: 12,07777