Cesium bromide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cesiumbromide
M-Clarity™ quality level = MQ100
429392-10G 429392-1G
SIAL429392-10GEA 183 EUR
SIAL429392-10G SIAL429392-1G
Cesium bromide, Sigma-Aldrich®
Cesium bromide
Formula: BrCs
MW: 212.81 g/mol
Boiling Pt: 1300
Melting Pt: 636
Density: 4.46 g/cm³ (20 °C)
MDL Number: MFCD00010954
CAS Number: 7787-69-1
EINECS: 232-130-0
Merck Index: 13,02019