Cesium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cesium Chloride
M-Clarity™ quality level = MQ100
449733-25G 449733-5G
SIAL449733-25GEA 393 EUR
SIAL449733-25G SIAL449733-5G
Cesium chloride, Sigma-Aldrich®
Cesium chloride
Formula: CsCl
Boiling Pt: 1303 °C (1013 hPa)
Melting Pt: 646 °C
Density: 3.7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021