Lithium chloride, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
203637-100G 203637-10G 203637-50G
SIAL203637-100GEA 559 EUR
SIAL203637-100G SIAL203637-10G SIAL203637-50G
Lithium chloride, Sigma-Aldrich®
Lithium chloride
Formula: LiCl
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550