Cesium carbonate ≥99.994% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
|
Warning
|
12117.22
12117.06
12117.14
12117.22EA
385
EUR
12117.22
12117.06
12117.14
Cesium carbonate ≥99.994% (metals basis), Puratronic®
Cesium carbonate
Purity: 99.994%
CAS No.: 534-17-8
Molecular Formula: Cs2CO3
Formula Weight: 325.82
Physical Form: Powder/Granules
Density: 4.072
pH: 10-12
Melting Point: 610° dec.
Beilstein Reference: 4546405
Merck Reference: 14,2010
MDL No.: MFCD00010957
Formula:
Cs₂CO₃ Melting Pt: 610 °C (dec.) Density: 4.072 Storage Temperature: Ambient |
MDL Number:
MFCD00010957 CAS Number: 534-17-8 EINECS: 208-591-9 Merck Index: 14,2010 |