Cesium carbonate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
20960-10G-F
20960-250G-F
20960-50G-F
SIAL20960-10G-FEA
32.5
EUR
SIAL20960-10G-F
SIAL20960-250G-F
SIAL20960-50G-F
Cesium carbonate, Sigma-Aldrich®
Cesium carbonate
Formula:
Cs₂CO₃ MW: 325.82 g/mol Melting Pt: 610 °C Density: 4.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010957 CAS Number: 534-17-8 EINECS: 208-591-9 Merck Index: 13,02020 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...