Potassium carbonate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Potash
M-Clarity™ quality level = MQ200
367877-10G
367877-50G
SIAL367877-10GEA
71.6
EUR
SIAL367877-10G
SIAL367877-50G
Potassium carbonate, Sigma-Aldrich®
Potassium carbonate
Formula:
K₂CO₃ Melting Pt: 891 °C Density: 2.42 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011382 CAS Number: 584-08-7 EINECS: 209-529-3 Merck Index: 13,07702 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...