Potassium carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Potash
MQ100
590681-25G 590681-5G
SIAL590681-25GEA 206 EUR
SIAL590681-25G SIAL590681-5G
Potassium carbonate, Sigma-Aldrich®
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891
Density: 2.42 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 13,07702
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5900faa1ba52c3213eed8f6bb2d952bac1f30f&vid=6298361525e6af416556b5694c74a8a91faa437c&prid=web&clid=SAPDAM