Lithium carbonate
Supplier: Roth Carl
|
Warning
|
3995.1
3995.2
ROTH3995.1EA
156
EUR
ROTH3995.1
ROTH3995.2
Lithium carbonate
Lithium carbonate
Formula:
Li₂CO₃ MW: 73.89 g/mol Melting Pt: 720 °C Density: 2.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 Merck Index: 13,05549 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...