Lithium carbonate, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
203629-10G 203629-50G
SIAL203629-10GEA 45.4 EUR
SIAL203629-10G SIAL203629-50G
Lithium carbonate, Sigma-Aldrich®
Lithium carbonate
Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549