Potassium dichromate, Sigma-Aldrich®
Supplier: Merck
Danger
MQ100
483044-10G 483044-50G
SIAL483044-10GEA 198 EUR
SIAL483044-10G SIAL483044-50G
Potassium dichromate, Sigma-Aldrich®
Potassium dichromate
Formula: K₂Cr₂O₇
MW: 294.18 g/mol
Boiling Pt: 500
Melting Pt: 398
Density: 2.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011367
CAS Number: 7778-50-9
EINECS: 231-906-6
UN: 3087
ADR: 5.1 II
REACH: 01-2119454792-32
Merck Index: 12,07790