Cesium fluoride 99.9%, extra pure
Supplier: Thermo Fisher Scientific
Danger
315910250 315910050 315911000
ACRO315910250EA 110 EUR
ACRO315910250 ACRO315910050 ACRO315911000
Cesium fluoride 99.9%, extra pure
Cesium fluoride
Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251
Melting Pt: 682 °C
Density: 4.627
Storage Temperature: Ambient
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 3288
ADR: 6.1 II
Order Now
Specification Test Results
Appearance White Powder
Total trace metal impurities ≤0.1 %