Cesium fluoride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

255718-10G 255718-50G
SIAL255718-10GEA 187 EUR
SIAL255718-10G SIAL255718-50G
Cesium fluoride, Sigma-Aldrich®
Cesium fluoride
Formula: CsF
Boiling Pt: 1251 °C (1013 hPa)
Melting Pt: 680 °C
Density: 4.627 g/cm³ (20 °C)
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR