Potassium fluoride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
MQ100
449148-25G
449148-5G
SIAL449148-25GEA
345
EUR
SIAL449148-25G
SIAL449148-5G
Potassium fluoride, Sigma-Aldrich®
Potassium fluoride
Formula:
KF MW: 58.1 g/mol Boiling Pt: 1505 Melting Pt: 860 Density: 2.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1 III Merck Index: 13,07714 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...