Potassium hexacyanoferrate(III)
Supplier: Roth Carl
Synonyms: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
7971.1 7971.2
ROTH7971.1EA 0 EUR
ROTH7971.1 ROTH7971.2
Potassium hexacyanoferrate(III)
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
MW: 329.25 g/mol
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
Merck Index: 13,07712